Anti-CES Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4669S 200-4669S
CA11029-088EA 258.98 CAD
CA11029-088 RL2004669S
Anti-CES Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CES
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR