Anti-LASNASE Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4671S 200-4671S
CA11029-090EA 258.98 CAD
RL2004671S CA11029-090
Anti-LASNASE Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: LASNASE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR