Anti-TRYP Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4680-0100 200-4680-0100
CA11029-330EA 949.55 CAD
RL20046800100 CA11029-330
Anti-TRYP Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: TRYP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR