Anti-AKT2 Rat Monoclonal Antibody [clone: 16G11.E8]

Supplier: Rockland Immunochemicals
200-501-E71L 200-501-E71L
CA11029-222EA 5491.83 CAD
CA11029-222 RL200501E71L
Anti-AKT2 Rat Monoclonal Antibody [clone: 16G11.E8]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 16G11.E8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR