Anti-ARHGAP22 PS22 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J11 600-401-J11
CA11029-126EA 858.79 CAD
RL600401J11 CA11029-126
Anti-ARHGAP22 PS22 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ARHGAP22 PS22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR