Anti-Muscarinic Acetylcholine Receptor 2 Rabbit Polyclonal Antibody

Supplier: Boster
PA1325-1
10205-892EA 1223.24 CAD
10205-892
Anti-Muscarinic Acetylcholine Receptor 2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Muscarinic acetylcholine receptor M2(CHRM2) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Muscarinic Acetylcholine Receptor 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR