Anti-Indoleamine 2, 3-dioxygenase Rabbit Polyclonal Antibody

Supplier: Boster
PA1611
10205-908EA 1223.24 CAD
10205-908
Anti-Indoleamine 2, 3-dioxygenase Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Indoleamine 2,3-dioxygenase 1(IDO1) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Indoleamine 2, 3-dioxygenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR