Anti-AHR Rabbit Polyclonal Antibody

Supplier: Boster
PA1782-2
10205-950EA 1223.24 CAD
10205-950
Anti-AHR Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Aryl hydrocarbon receptor(AHR) detection. Tested with WB, IHC-P in Mouse.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR