Anti-CALB1 Mouse Monoclonal Antibody [clone: CB-D7]

Supplier: Boster
MA1007
10205-958EA 1222.9 CAD
10205-958
Anti-CALB1 Mouse Monoclonal Antibody [clone: CB-D7]
Antibodies
Mouse IgG monoclonal antibody for Calbindin-D, calbindin 1, 28kDa (CALB1) detection. Tested with WB, IHC-P in Human;mouse;rat;bovine. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: CALB1
Clonality: Monoclonal
Clone: CB-D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR