Anti-FN1 Mouse Monoclonal Antibody [clone: FN3E2]

Supplier: Boster
MA1116
10205-966EA 1222.9 CAD
10205-966
Anti-FN1 Mouse Monoclonal Antibody [clone: FN3E2]
Antibodies
Mouse IgG monoclonal antibody for Fibronectin, fibronectin 1 (FN1 ) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;mouse;rat;chicken. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Fibronectin
Clonality: monoclonal
Clone: FN3E2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR