Anti-GABRA1 Rabbit Polyclonal Antibody

Supplier: Boster
PA1578
10205-986EA 1223.24 CAD
10205-986
Anti-GABRA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Gamma-aminobutyric acid receptor subunit alpha-1(GABRA1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: GABA A Receptor alpha 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR