Anti-MB Mouse Monoclonal Antibody [clone: 42381]

Supplier: Boster
MA1062
10205-994EA 1223.24 CAD
10205-994
Anti-MB Mouse Monoclonal Antibody [clone: 42381]
Antibodies
Mouse IgG monoclonal antibody for Myoglobin, myoglobin (MB) detection. Tested with WB, IHC-P in Human;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: MB
Clonality: Monoclonal
Clone: 42381
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR