Anti-Filensin Mouse Monoclonal Antibody [clone: FIL-27]

Supplier: Boster Biological Technology
MA1041
10206-020EA 1223.24 CAD
10206-020
Anti-Filensin Mouse Monoclonal Antibody [clone: FIL-27]
Antibodies
Mouse IgG monoclonal antibody for Filensin, beaded filament structural protein 1, filensin (BFSP1) detection. Tested with WB, IHC-F in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Filensin
Clonality: monoclonal
Clone: FIL-27
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR