Anti-p19INK4d Mouse Monoclonal Antibody [clone: IMD-19]

Supplier: Boster
MA1075
10206-026EA 1223.24 CAD
10206-026
Anti-p19INK4d Mouse Monoclonal Antibody [clone: IMD-19]
Antibodies
Mouse IgG monoclonal antibody for p19INK4d, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4) (CDKN2D) detection. Tested with WB, IHC-P, ICC in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: p19INK4d
Clonality: Monoclonal
Clone: IMD-19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR