Anti-VIM Mouse Monoclonal Antibody [clone: VMT-24]

Supplier: Boster
MA1102
10206-032EA 1223.24 CAD
10206-032
Anti-VIM Mouse Monoclonal Antibody [clone: VMT-24]
Antibodies
Mouse IgG monoclonal antibody for Vimentin, vimentin (VIM) detection. Tested with WB, IHC-P in Human;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: VMT-24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR