Anti-VDR Rabbit Polyclonal Antibody

Supplier: Boster
PA1983
10206-144EA 1222.89 CAD
10206-144
Anti-VDR Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Vitamin D3 receptor(VDR) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: VDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR