Anti-CDK6 Mouse Monoclonal Antibody [clone: IML-6]

Supplier: Boster
MA1021
10206-158EA 1223.24 CAD
10206-158
Anti-CDK6 Mouse Monoclonal Antibody [clone: IML-6]
Antibodies
Mouse IgG monoclonal antibody for CDK6, cyclin-dependent kinase 6 (CDK6) detection. Tested with WB, ICC in Human;mouse;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: CDK6
Clonality: Monoclonal
Clone: IML-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR