Anti-CD4 Mouse Monoclonal Antibody [clone: CA-4]

Supplier: Boster
MA1016
10206-190EA 1223.24 CAD
10206-190
Anti-CD4 Mouse Monoclonal Antibody [clone: CA-4]
Antibodies
Mouse IgG monoclonal antibody for CD4, CD4 molecule (CD4) detection. Tested with IHC-F in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: CA-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR