Anti-MAPT Mouse Monoclonal Antibody [clone: TAU-93]

Supplier: Boster
MA1093
10206-200EA 1223.24 CAD
10206-200
Anti-MAPT Mouse Monoclonal Antibody [clone: TAU-93]
Antibodies
Mouse IgG monoclonal antibody for Tau, microtubule-associated protein tau (MAPT) detection. Tested with WB, IHC-P in Human;mouse;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: MAPT
Clonality: Monoclonal
Clone: TAU-93
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR