Anti-AGT Rabbit Polyclonal Antibody

Supplier: Boster
PA1643
10206-262EA 1221.97 CAD
10206-262
Anti-AGT Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Angiotensinogen(AGT) detection. Tested with WB in Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR