Anti-Catenin gamma (Plakoglobin) Mouse Monoclonal Antibody [clone: CN-3]

Supplier: Boster
MA1012
10206-308EA 1223.24 CAD
10206-308
Anti-Catenin gamma (Plakoglobin) Mouse Monoclonal Antibody [clone: CN-3]
Antibodies
Mouse IgG monoclonal antibody for Catenin gamma (Plakoglobin), junction plakoglobin ( JUP) detection. Tested with WB, IHC-F, ICC in Human;chicken;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: Catenin gamma (Plakoglobin)
Clonality: monoclonal
Clone: CN-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR