Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
RP1026
10206-310EA 1223.16 CAD
10206-310
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Baculoviral IAP repeat-containing protein 5(BIRC5) detection. Tested with WB, IHC-P, ELISA in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR