Anti-OSM Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1668
10206-356EA 1221.94 CAD
10206-356
Anti-OSM Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Oncostatin-M(OSM) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: OSM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR