Anti-KLK9 Rabbit Polyclonal Antibody

Supplier: Boster
PA1820
10206-366EA 1221.97 CAD
10206-366
Anti-KLK9 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Kallikrein-9(KLK9) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: KLK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR