Anti-CCND1 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1245-1
10206-464EA 1221.97 CAD
10206-464
Anti-CCND1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for G1/S-specific cyclin-D1(CCND1) detection. Tested with WB in Human;Mouse;Rat.

  • Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: CCND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR