Anti-GRP78 BiP Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1815
10206-500EA 1226.32 CAD
10206-500
Anti-GRP78 BiP Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for 78 kDa glucose-regulated protein(HSPA5) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: GRP78 BiP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR