Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SELENBP1 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology

Rabbit IgG polyclonal antibody for Selenium-binding protein 1(SELENBP1) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: SELENBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg PA2216 10207-912 Each Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Amino acid number 226 to 240
Antibody type Primary
Antigen name Selenium Binding Protein 1
Antigen symbol SELENBP1
Antigen synonyms SBP1, SBP, Lp56, Selenium binding protein2, SBP 1, Lp 56, SP56, Selenium binding protein1, SELNBP1, Selenbp2, hSBP, SP 56, Selenium binding protein 2, 56 a selenium binding protein, SELENBP 1, Selen bp2, Lpsb, SBP56, Selenbp 2
Clonality Polyclonal
Conjugation Unconjugated
Cross Adsorption No
Form Lyophilized
Host Rabbit
ImmunoChemistry Yes
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Selenium Binding Protein 1 (226-240aa SHLYVWDWQRHEIVQ).
Isotype IgG
Purification purified by immunogen affinity purification
Reactivity Human
Sequence SHLYVWDWQRHEIVQ
Storage temperature -20°C
Western Blot Yes
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty