Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-5HT2A Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
PA1373
Rabbit IgG polyclonal antibody for 5-hydroxytryptamine receptor 2A(HTR2A) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: 5HT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg PA1373 10207-988 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol 5HT2A
Antigen name 5-hydroxytryptamine receptor 2A
Reactivity Mouse, Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms 5-HT-2A, 5-HT2A, 5-HT-2, HTR2, HTR2A, Serotonin receptor 2A
Amino acid number 418 to 432
Epitope near the C-terminus
Cross adsorption No
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human 5HT2A Receptor (418-432aa AYKSSQLQMGQKKNS), different from the mouse sequence by one amino acid.
Purification purified by immunogen affinity purification
Sequence AYKSSQLQMGQKKNS
Form Lyophilized
Storage temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty