Anti-BTG4 Mouse Monoclonal Antibody [clone: 1A6]

Supplier: Abnova
H00054766-M07
10578-152EA 502.5 CAD
10578-152
Anti-BTG4 Mouse Monoclonal Antibody [clone: 1A6]
Antibodies
Type: Primary
Antigen: BTG4
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR