Anti-KCTD9 Mouse Monoclonal Antibody [clone: 1F3]

Supplier: Abnova
H00054793-M01
10586-216EA 502.5 CAD
10586-216
Anti-KCTD9 Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Type: Primary
Antigen: KCTD9
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR