Anti-AHI1 Mouse Polyclonal Antibody

H00054806-B01P
10510-544EA 420.99 CAD
10510-544
Anti-AHI1 Mouse Polyclonal Antibody
Type: Primary
Antigen: AHI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR