Anti-GIPC2 Mouse Polyclonal Antibody

H00054810-B01P
10557-492EA 420.99 CAD
10557-492
Anti-GIPC2 Mouse Polyclonal Antibody
Type: Primary
Antigen: GIPC2 (GIPC PDZ domain containing family, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR