Anti-NDE1 Mouse Monoclonal Antibody [clone: 2G11-1C11]

Supplier: Abnova
H00054820-M01
10594-238EA 502.5 CAD
10594-238
Anti-NDE1 Mouse Monoclonal Antibody [clone: 2G11-1C11]
Antibodies
Type: Primary
Antigen: NDE1
Clonality: Monoclonal
Clone: 2G11-1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR