Anti-ZCCHC2 Mouse Polyclonal Antibody

Supplier: Abnova
H00054877-B01P
10574-170EA 502.5 CAD
10574-170
Anti-ZCCHC2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZCCHC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR