Anti-ODAM Mouse Polyclonal Antibody

Supplier: Abnova
H00054959-B01P
10566-278EA 502.5 CAD
10566-278
Anti-ODAM Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ODAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR