Anti-THG1L Mouse Polyclonal Antibody

H00054974-B01P
10561-270EA 420.99 CAD
10561-270
Anti-THG1L Mouse Polyclonal Antibody
Type: Primary
Antigen: THG1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR