Anti-THG1L Mouse Monoclonal Antibody [clone: 4F11]

Supplier: Abnova
H00054974-M01
10589-696EA 502.5 CAD
10589-696
Anti-THG1L Mouse Monoclonal Antibody [clone: 4F11]
Antibodies
Type: Primary
Antigen: THG1L
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR