Anti-ULK4 Mouse Monoclonal Antibody [clone: 4A10-1A7]

Supplier: Abnova
H00054986-M01
10586-228EA 502.5 CAD
10586-228
Anti-ULK4 Mouse Monoclonal Antibody [clone: 4A10-1A7]
Antibodies
Type: Primary
Antigen: ULK4
Clonality: Monoclonal
Clone: 4A10-1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR