Anti-PIH1D1 Mouse Monoclonal Antibody [clone: 1000000]

Supplier: Abnova
H00055011-M05
10586-236EA 502.5 CAD
10586-236
Anti-PIH1D1 Mouse Monoclonal Antibody [clone: 1000000]
Antibodies
Type: Primary
Antigen: PIH1D1
Clonality: Monoclonal
Clone: 1000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR