Anti-TTC38 Mouse Polyclonal Antibody

H00055020-B01
10560-726EA 392.79 CAD
10560-726
Anti-TTC38 Mouse Polyclonal Antibody
Type: Primary
Antigen: TTC38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR