Anti-RALGPS2 Mouse Monoclonal Antibody [clone: 3F1]

Supplier: Abnova
H00055103-M01
10599-118EA 502.5 CAD
10599-118
Anti-RALGPS2 Mouse Monoclonal Antibody [clone: 3F1]
Antibodies
Type: Primary
Antigen: RALGPS2
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR