Anti-FANCL Rabbit Polyclonal Antibody

Supplier: Abnova
H00055120-D01
10585-656EA 564 CAD
10585-656
Anti-FANCL Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FANCL (Fanconi anemia, complementation group L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR