Anti-ARMC4 Mouse Monoclonal Antibody [clone: 5F1]

Supplier: Abnova
H00055130-M02A
10621-242EA 502.5 CAD
10621-242
Anti-ARMC4 Mouse Monoclonal Antibody [clone: 5F1]
Antibodies
Type: Primary
Antigen: ARMC4
Clonality: Monoclonal
Clone: 5F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR