Anti-CDCA8 Rabbit Polyclonal Antibody

Supplier: Abnova
H00055143-D01P
10581-678EA 564 CAD
10581-678
Anti-CDCA8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDCA8
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR