Anti-RFWD3 Mouse Monoclonal Antibody [clone: 6B4]

Supplier: Abnova
H00055159-M01
10599-620EA 502.5 CAD
10599-620
Anti-RFWD3 Mouse Monoclonal Antibody [clone: 6B4]
Antibodies
Type: Primary
Antigen: RFWD3
Clonality: Monoclonal
Clone: 6B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR