Anti-RNF220 Mouse Monoclonal Antibody [clone: 500000]

Supplier: Abnova
H00055182-M01
10586-140EA 502.5 CAD
10586-140
Anti-RNF220 Mouse Monoclonal Antibody [clone: 500000]
Antibodies
Type: Primary
Antigen: RNF220
Clonality: Monoclonal
Clone: 500000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR