Anti-PBRM1 (polybromo 1) Mouse Polyclonal Antibody

Supplier: Abnova
H00055193-B02
10509-756EA 502.5 CAD
10509-756
Anti-PBRM1 (polybromo 1) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PBRM1 (polybromo 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR