Anti-MAP1S Mouse Monoclonal Antibody [clone: 4H2]

Supplier: Abnova
H00055201-M05
10578-058EA 502.5 CAD
10578-058
Anti-MAP1S Mouse Monoclonal Antibody [clone: 4H2]
Antibodies
Type: Primary
Antigen: MAP1S
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR