Anti-EXD2 Mouse Polyclonal Antibody

H00055218-B01P
10556-126EA 420.99 CAD
10556-126
Anti-EXD2 Mouse Polyclonal Antibody
Type: Primary
Antigen: EXD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR