Anti-FGGY Mouse Monoclonal Antibody [clone: 3B9]

Supplier: Abnova
H00055277-M04
10586-148EA 502.5 CAD
10586-148
Anti-FGGY Mouse Monoclonal Antibody [clone: 3B9]
Antibodies
Type: Primary
Antigen: FGGY (FGGY carbohydrate kinase domain containing)
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR